Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc77
Entrez Gene ID
67200
Gene Name
coiled-coil domain containing 77
Gene Aliases
2400002C23Rik, 2700091N06Rik, AV168274
Chromosome Location
Chr.6: 120324322 - 120364369 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026028.5 NP_080304.1 7 - 8 835 87
NR_045558.1 6 - 7 858 87
GenBank mRNA AK012598.1 - 7 - 8 727 87
AK031590.1 - 6 - 7 859 87
BC018387.1 - 7 - 8 663 87

View Full Product Details