Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbln2
Entrez Gene ID
14115
Gene Name
fibulin 2
Gene Aliases
5730577E14Rik, FIBL-2
Chromosome Location
Chr.6: 91212764 - 91272540 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081437.1 NP_001074906.1 11 - 12 2687 70
NM_007992.2 NP_032018.2 12 - 13 2828 70
GenBank mRNA AK159361.1 - 11 - 12 2999 70
AK161522.1 - 11 - 12 2630 70
AK168688.1 - 11 - 12 2625 70
BC005443.1 - 11 - 12 2667 70
X75285.1 - 12 - 13 2728 70

View Full Product Details