The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081437.1 | NP_001074906.1 | 16 - 17 | 3316 | 65 |
NM_007992.2 | NP_032018.2 | 17 - 18 | 3457 | 65 | |
GenBank mRNA | AK159361.1 | - | 16 - 17 | 3628 | 65 |
AK161522.1 | - | 16 - 17 | 3259 | 65 | |
AK168688.1 | - | 16 - 17 | 3254 | 65 | |
BC005443.1 | - | 16 - 17 | 3296 | 65 |