The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177556.1 | NP_001171027.1 | 3 - 4 | 300 | 87 |
NM_001177557.1 | NP_001171028.1 | 3 - 4 | 312 | 87 | |
NM_001177558.1 | NP_001171029.1 | 4 - 5 | 333 | 87 | |
NM_001177559.1 | NP_001171030.1 | 4 - 5 | 353 | 87 | |
NM_001177560.1 | NP_001171031.1 | 4 - 5 | 370 | 87 | |
NM_025278.5 | NP_079554.1 | 4 - 5 | 365 | 87 | |
GenBank mRNA | AK005561.1 | - | 3 - 4 | 286 | 87 |
AK033422.1 | - | 3 - 4 | 224 | 87 | |
AK148765.1 | - | 3 - 4 | 250 | 87 | |
AK159872.1 | - | 4 - 5 | 307 | 87 | |
AK162342.1 | - | 4 - 5 | 333 | 87 | |
AK166035.1 | - | 3 - 4 | 257 | 87 | |
AK166299.1 | - | 4 - 5 | 333 | 87 | |
BB851888.1 | - | 4 - 5 | 366 | 87 | |
BC040685.1 | - | 4 - 5 | 331 | 87 |