Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekhg1
Entrez Gene ID
213783
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 1
Gene Aliases
D10Ertd733e, Gm521, mKIAA1209
Chromosome Location
Chr.10: 3740251 - 3967303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033253.3 NP_001028425.1 11 - 12 1481 70
NM_001159942.1 NP_001153414.1 12 - 13 1647 70
GenBank mRNA AK122468.1 - 8 - 9 805 70
AK135923.1 - 11 - 12 1483 70
BC076628.1 - 9 - 10 878 70
BC117965.1 - 10 - 11 1083 70
BC117966.1 - 10 - 11 1076 70

View Full Product Details