Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec4a1
Entrez Gene ID
269799
Gene Name
C-type lectin domain family 4, member a1
Gene Aliases
BC049354, Dcir4
Chromosome Location
Chr.6: 122921848 - 122934619 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199311.2 NP_955015.1 3 - 4 443 116
GenBank mRNA AY397675.1 - 3 - 4 378 116
BC049354.1 - 3 - 4 433 116
CK626014.1 - 3 - 4 443 116

View Full Product Details