The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199311.2 | NP_955015.1 | 5 - 6 | 711 | 64 |
GenBank mRNA | AY397675.1 | - | 5 - 6 | 646 | 64 |
BC049354.1 | - | 5 - 6 | 701 | 64 |