The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023547.1 | NP_076036.1 | 4 - 5 | 497 | 60 |
GenBank mRNA | AK010946.1 | - | 4 - 5 | 599 | 60 |
BC019383.1 | - | 4 - 5 | 531 | 60 | |
BC047990.1 | - | 4 - 5 | 532 | 60 | |
BC058788.1 | - | 4 - 5 | 543 | 60 |