Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lag3
Entrez Gene ID
16768
Gene Name
lymphocyte-activation gene 3
Gene Aliases
CD223, LAG-3, Ly66
Chromosome Location
Chr.6: 124904359 - 124912435 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008479.2 NP_032505.1 7 - 8 1760 111
GenBank mRNA BC120591.1 - 7 - 8 1692 111
X98113.1 - 7 - 8 1762 111

View Full Product Details