Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn1r67
Entrez Gene ID
171263
Gene Name
vomeronasal 1 receptor 67
Gene Aliases
V1re10
Chromosome Location
Chr.7: 10446789 - 10447787 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134229.2 NP_598990.2 1 - 1 896 75
GenBank mRNA BC138174.1 - 1 - 1 896 75
BC145742.1 - 1 - 1 896 75

View Full Product Details