Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lace1
Entrez Gene ID
215951
Gene Name
lactation elevated 1
Gene Aliases
CG8520
Chromosome Location
Chr.10: 42312581 - 42478584 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145743.2 NP_665686.2 7 - 8 966 66
GenBank mRNA AF397909.1 - 6 - 7 669 66
AF520417.1 - 7 - 8 815 66
AK044595.1 - 7 - 8 842 66
BC023308.1 - 6 - 7 643 66
BC089595.1 - 7 - 8 816 66

View Full Product Details