Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aim1
Entrez Gene ID
11630
Gene Name
absent in melanoma 1
Gene Aliases
AI462491, AI463325
Chromosome Location
Chr.10: 43950307 - 44148924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172393.2 NP_765981.2 19 - 20 5770 106
GenBank mRNA AF397913.1 - 13 - 14 1478 106
AK041082.1 - 18 - 19 2387 106
AK088102.1 - 18 - 19 2442 106
BC035551.1 - 18 - 19 2922 106
BC059272.1 - 18 - 19 2927 106

View Full Product Details