Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep57l1
Entrez Gene ID
103268
Gene Name
centrosomal protein 57-like 1
Gene Aliases
2410017P07Rik, 4930484D11Rik, AI607327, cep57R
Chromosome Location
Chr.10: 41718840 - 41809868 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243074.1 NP_001230003.1 4 - 5 505 156
NM_026643.5 NP_080919.3 4 - 5 423 156
NM_029132.2 NP_083408.1 4 - 5 491 156
GenBank mRNA AK010530.1 - 4 - 5 442 156
AK015619.1 - 3 - 4 406 156
BC021375.1 - 4 - 5 515 156
BC138230.1 - 4 - 5 426 156
BQ553570.1 - 3 - 4 380 156
CD354087.1 - 1 - 2 173 156

View Full Product Details