Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txlna
Entrez Gene ID
109658
Gene Name
taxilin alpha
Gene Aliases
2600010N21Rik, AI425952, AW209241, Il14, Txln
Chromosome Location
Chr.4: 129626077 - 129641084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005506.3 NP_001005506.2 3 - 4 974 64
NM_001199695.1 NP_001186624.1 4 - 5 691 64
GenBank mRNA AK135290.1 - 3 - 4 822 64
AK141252.1 - 4 - 5 625 64
AK150240.1 - 4 - 5 648 64
AK160829.1 - 3 - 4 822 64
AK161649.1 - 4 - 5 654 64
AK168418.1 - 3 - 4 975 64
AK171101.1 - 4 - 5 667 64
BC060227.1 - 3 - 4 809 64
BC065115.1 - 3 - 4 809 64

View Full Product Details