Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kpna6
Entrez Gene ID
16650
Gene Name
karyopherin (importin) alpha 6
Gene Aliases
IPOA7, Kpna5, NPI-2
Chromosome Location
Chr.4: 129643979 - 129672767 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008468.4 NP_032494.3 5 - 6 519 86
GenBank mRNA AF020773.1 - 4 - 5 423 86
AK046371.1 - 5 - 6 478 86
AK084212.1 - 5 - 6 477 86
AK154882.1 - 5 - 6 504 86
BC004833.1 - 4 - 5 435 86
CT010260.1 - 4 - 5 423 86

View Full Product Details