Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ap1b1
Entrez Gene ID
11764
Gene Name
adaptor protein complex AP-1, beta 1 subunit
Gene Aliases
Adtb1, b2b1660Clo
Chromosome Location
Chr.11: 4947521 - 5042794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243043.1 NP_001229972.1 16 - 17 2630 67
NM_001243044.1 NP_001229973.1 15 - 16 2412 67
NM_007454.3 NP_031480.2 15 - 16 2412 67
GenBank mRNA AA003413.1 - 1 - 2 48 67
AK034140.1 - 16 - 17 2633 67
AK155892.1 - 15 - 16 2332 67
AK159273.1 - 15 - 16 2303 67
AK160042.1 - 15 - 16 2303 67
AK165747.1 - 15 - 16 2329 67
AK170376.1 - 15 - 16 2323 67
BC008513.1 - 15 - 16 2225 67
BC066827.1 - 6 - 7 956 67
Y07919.1 - 15 - 16 2267 67

View Full Product Details