Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ap1b1
Entrez Gene ID
11764
Gene Name
adaptor protein complex AP-1, beta 1 subunit
Gene Aliases
Adtb1, b2b1660Clo
Chromosome Location
Chr.11: 4947521 - 5042794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243043.1 NP_001229972.1 10 - 11 1719 83
NM_001243044.1 NP_001229973.1 10 - 11 1522 83
NM_007454.3 NP_031480.2 10 - 11 1522 83
GenBank mRNA AK034140.1 - 10 - 11 1722 83
AK155892.1 - 10 - 11 1442 83
AK159273.1 - 10 - 11 1413 83
AK160042.1 - 10 - 11 1413 83
AK165747.1 - 10 - 11 1439 83
AK170376.1 - 10 - 11 1433 83
BC008513.1 - 10 - 11 1335 83
BC066827.1 - 1 - 2 66 83
Y07919.1 - 10 - 11 1377 83

View Full Product Details