Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr92
Entrez Gene ID
103784
Gene Name
WD repeat domain 92
Gene Aliases
AI553587, HZGJ
Chromosome Location
Chr.11: 17211893 - 17235200 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178909.4 NP_849240.1 6 - 7 843 59
GenBank mRNA AK033294.1 - 6 - 7 789 59
AK047691.1 - 6 - 7 817 59
AK088218.1 - 6 - 7 818 59
AK214428.1 - 1 - 2 126 59
BC096540.1 - 6 - 7 772 59

View Full Product Details