Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myb
Entrez Gene ID
17863
Gene Name
myeloblastosis oncogene
Gene Aliases
AI550390, M16449, c-myb
Chromosome Location
Chr.10: 21124930 - 21160984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198914.1 NP_001185843.1 11 - 12 1951 76
NM_010848.3 NP_034978.3 10 - 11 1594 76
GenBank mRNA AK088079.1 - 11 - 12 1872 76
AK132250.1 - 10 - 11 1459 76
AK137794.1 - 4 - 5 539 76
AK137965.1 - 10 - 11 1460 76
BC011513.1 - 10 - 11 1502 76
M12848.1 - 10 - 11 1592 76
S83053.1 - 1 - 2 125 76
X02774.1 - 10 - 11 1364 76

View Full Product Details