Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myb
Entrez Gene ID
17863
Gene Name
myeloblastosis oncogene
Gene Aliases
AI550390, M16449, c-myb
Chromosome Location
Chr.10: 21124930 - 21160984 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198914.1 NP_001185843.1 12 - 13 2072 87
NM_010848.3 NP_034978.3 11 - 12 1715 87
GenBank mRNA AK088079.1 - 12 - 13 1993 87
AK132250.1 - 11 - 12 1580 87
AK137794.1 - 5 - 6 660 87
AK137965.1 - 11 - 12 1581 87
BC011513.1 - 11 - 12 1623 87
M12848.1 - 11 - 12 1713 87
S83053.1 - 2 - 3 246 87
X02774.1 - 11 - 12 1485 87

View Full Product Details