Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aig1
Entrez Gene ID
66253
Gene Name
androgen-induced 1
Gene Aliases
1500031O19Rik, AV064870, AW413422, CGI-103
Chromosome Location
Chr.10: 13647054 - 13869036 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK162306.1 - 5 - 6 772 93
BC035305.1 - 5 - 6 903 93
EF410113.1 - 3 - 4 632 93

View Full Product Details