The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153177.3 | NP_694817.2 | 7 - 8 | 1097 | 96 |
GenBank mRNA | AK030018.1 | - | 7 - 8 | 1113 | 96 |
AK129395.2 | - | 4 - 5 | 413 | 96 | |
BC096023.1 | - | 7 - 8 | 879 | 96 |