Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbln2
Entrez Gene ID
14115
Gene Name
fibulin 2
Gene Aliases
5730577E14Rik, FIBL-2
Chromosome Location
Chr.6: 91212764 - 91272540 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081437.1 NP_001074906.1 8 - 9 2278 116
GenBank mRNA AK159361.1 - 8 - 9 2590 116
AK161522.1 - 8 - 9 2221 116
AK168688.1 - 8 - 9 2216 116
BC005443.1 - 8 - 9 2258 116

View Full Product Details