Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tada3
Entrez Gene ID
101206
Gene Name
transcriptional adaptor 3
Gene Aliases
1110004B19Rik, ADA3, AI987856, Tada3l
Chromosome Location
Chr.6: 113366633 - 113377939 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133932.2 NP_598693.1 1 - 2 204 72
GenBank mRNA AK078906.1 - 1 - 2 524 72
BC026630.1 - 1 - 2 204 72

View Full Product Details