The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011763.2 | NP_035893.1 | 4 - 5 | 408 | 95 |
GenBank mRNA | AK007823.1 | - | 4 - 5 | 387 | 95 |
AK029052.1 | - | 4 - 5 | 394 | 95 | |
BC059273.1 | - | 3 - 4 | 272 | 95 |