The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138592.4 | NP_613058.1 | 12 - 13 | 1676 | 64 |
GenBank mRNA | AK010443.1 | - | 12 - 13 | 1686 | 64 |
AK018595.1 | - | 12 - 13 | 1677 | 64 | |
AK033105.1 | - | 12 - 13 | 1673 | 64 | |
AK045062.1 | - | 12 - 13 | 1673 | 64 | |
AK167668.1 | - | 12 - 13 | 1664 | 64 | |
AK191104.1 | - | 1 - 2 | 103 | 64 | |
AK216737.1 | - | 1 - 2 | 103 | 64 | |
BC026983.1 | - | 12 - 13 | 1651 | 64 |