Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prrt3
Entrez Gene ID
210673
Gene Name
proline-rich transmembrane protein 3
Gene Aliases
6330505P20, B230206N24Rik
Chromosome Location
Chr.6: 113493638 - 113502235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289699.1 NP_001276628.1 1 - 2 78 72
GenBank mRNA AK045678.1 - 1 - 2 40 72

View Full Product Details