Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vip
Entrez Gene ID
22353
Gene Name
vasoactive intestinal polypeptide
Gene Aliases
-
Chromosome Location
Chr.10: 5639193 - 5647617 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011702.2 NP_035832.1 1 - 2 154 57
GenBank mRNA AK018599.1 - 1 - 2 155 57
BC089511.1 - 1 - 2 60 57

View Full Product Details