Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Col10a1
Entrez Gene ID
12813
Gene Name
collagen, type X, alpha 1
Gene Aliases
Col10, Col10a-1
Chromosome Location
Chr.10: 34389981 - 34397085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009925.4 NP_034055.1 1 - 2 60 66
GenBank mRNA BY141606.1 - 1 - 2 61 66

View Full Product Details