Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zmynd12
Entrez Gene ID
332934
Gene Name
zinc finger, MYND domain containing 12
Gene Aliases
AI414736
Chromosome Location
Chr.4: 119422657 - 119453899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014900.2 NP_001014900.2 6 - 7 1033 91
GenBank mRNA AY959941.1 - 6 - 7 844 91
BC147552.1 - 6 - 7 943 91
BC147553.1 - 6 - 7 943 91
BC147848.1 - 6 - 7 1033 91
BC147851.1 - 6 - 7 1033 91

View Full Product Details