The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008190.1 | NP_032216.1 | 2 - 3 | 301 | 74 |
GenBank mRNA | AK075814.1 | - | 2 - 3 | 320 | 74 |
BC012640.1 | - | 2 - 3 | 312 | 74 | |
L05516.1 | - | 2 - 3 | 295 | 74 | |
M95175.1 | - | 2 - 3 | 301 | 74 |