Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Edn2
Entrez Gene ID
13615
Gene Name
endothelin 2
Gene Aliases
VIC
Chromosome Location
Chr.4: 120161424 - 120167360 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007902.2 NP_031928.2 2 - 3 298 73
GenBank mRNA BC037042.1 - 2 - 3 527 73
X59556.1 - 2 - 3 289 73

View Full Product Details