The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009566.4 | NP_033592.2 | 4 - 5 | 322 | 72 |
GenBank mRNA | AK141522.1 | - | 4 - 5 | 323 | 72 |
BC096436.1 | - | 3 - 4 | 373 | 72 | |
U47104.1 | - | 4 - 5 | 342 | 72 |