Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt8b
Entrez Gene ID
22423
Gene Name
wingless-type MMTV integration site family, member 8B
Gene Aliases
-
Chromosome Location
Chr.19: 44493472 - 44514273 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC119544.2 - 4 - 5 432 78

View Full Product Details