The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112701.1 | NP_001106171.1 | 1 - 2 | 213 | 82 |
NM_011181.3 | NP_035311.1 | 1 - 2 | 213 | 82 | |
GenBank mRNA | AB013466.1 | - | 1 - 2 | 148 | 82 |
AF079971.1 | - | 1 - 2 | 182 | 82 | |
AK045451.1 | - | 1 - 2 | 182 | 82 | |
AK078067.1 | - | 1 - 2 | 210 | 82 | |
AK143829.1 | - | 1 - 2 | 155 | 82 | |
AK168979.1 | - | 1 - 2 | 153 | 82 | |
AK172050.1 | - | 1 - 2 | 169 | 82 | |
BC004662.1 | - | 1 - 2 | 127 | 82 | |
BC088833.1 | - | 1 - 2 | 187 | 82 |