Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyth2
Entrez Gene ID
19158
Gene Name
cytohesin 2
Gene Aliases
ARNO, CLM2, Pscd2
Chromosome Location
Chr.7: 45806637 - 45814316 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112701.1 NP_001106171.1 6 - 7 745 74
NM_011181.3 NP_035311.1 6 - 7 745 74
GenBank mRNA AB013466.1 - 6 - 7 680 74
AB013467.1 - 5 - 6 1248 74
AF079971.1 - 6 - 7 714 74
AK045451.1 - 6 - 7 714 74
AK143829.1 - 6 - 7 687 74
AK168979.1 - 6 - 7 685 74
AK172050.1 - 6 - 7 701 74
BC004662.1 - 6 - 7 659 74
BC088833.1 - 6 - 7 719 74

View Full Product Details