The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016860.1 | NP_058556.1 | 10 - 11 | 1082 | 66 |
GenBank mRNA | AB010297.1 | - | 10 - 11 | 1082 | 66 |
AK010632.1 | - | 10 - 11 | 1079 | 66 | |
AK019518.1 | - | 9 - 10 | 1024 | 66 | |
AK088062.1 | - | 10 - 11 | 1089 | 66 | |
AK162248.1 | - | 10 - 11 | 1098 | 66 | |
AK167450.1 | - | 10 - 11 | 1089 | 66 | |
AK168889.1 | - | 10 - 11 | 1086 | 66 | |
AK214590.1 | - | 1 - 2 | 50 | 66 | |
BC007131.1 | - | 10 - 11 | 1097 | 66 |