The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161515.1 | NP_001154987.1 | 2 - 3 | 403 | 101 |
NM_001161516.1 | NP_001154988.1 | 2 - 3 | 184 | 101 | |
NM_178788.4 | NP_848903.2 | 2 - 3 | 425 | 101 | |
NR_027759.1 | 2 - 3 | 425 | 101 | ||
GenBank mRNA | AK031645.1 | - | 2 - 3 | 390 | 101 |
AK039066.1 | - | 2 - 3 | 215 | 101 | |
AK138614.1 | - | 2 - 3 | 213 | 101 | |
AK171894.1 | - | 2 - 3 | 222 | 101 | |
BB621273.1 | - | 2 - 3 | 390 | 101 | |
BC031719.1 | - | 2 - 3 | 209 | 101 | |
CO803572.1 | - | 2 - 3 | 184 | 101 |