The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021392.4 | NP_067367.3 | 3 - 4 | 419 | 66 |
GenBank mRNA | AF242858.1 | - | 3 - 4 | 287 | 66 |
AK031638.1 | - | 3 - 4 | 422 | 66 | |
AK165187.1 | - | 2 - 3 | 655 | 66 | |
BC011174.1 | - | 3 - 4 | 304 | 66 |