Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxk1
Entrez Gene ID
17425
Gene Name
forkhead box K1
Gene Aliases
A630048H08Rik, AI463295, ENSMUSG00000075577, Gm10868, Mnf
Chromosome Location
Chr.5: 142401497 - 142462015 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199068.2 NP_951031.2 8 - 9 1910 101
GenBank mRNA AK144418.1 - 8 - 9 1803 101
AK147375.1 - 8 - 9 1908 101
BC002273.1 - 2 - 3 345 101
BC060238.1 - 8 - 9 1888 101
L26507.1 - 8 - 9 1887 101

View Full Product Details