Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dennd3
Entrez Gene ID
105841
Gene Name
DENN/MADD domain containing 3
Gene Aliases
AI447457, E030003N15Rik
Chromosome Location
Chr.15: 73512560 - 73572242 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081066.1 NP_001074535.1 7 - 8 1270 73
GenBank mRNA AK173046.1 - 7 - 8 1170 73
BC132389.1 - 7 - 8 1158 73
BC137791.1 - 7 - 8 1158 73
BC144912.1 - 7 - 8 1158 73

View Full Product Details