Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc1a1
- Entrez Gene ID
-
20510
- Gene Name
- solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1
- Gene Aliases
-
D130048G10Rik, EAAC1, EAAC2, EAAT3, MEAAC1
- Chromosome Location
-
Chr.19: 28835166 - 28913960 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF087578.1 |
- |
|
|
|
Chr. - - [-] on Build |
65 |
View Full Product Details