Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ide
Entrez Gene ID
15925
Gene Name
insulin degrading enzyme
Gene Aliases
1300012G03Rik, 4833415K22Rik, AA675336, AI507533
Chromosome Location
Chr.19: 37268741 - 37334544 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031156.3 NP_112419.3 4 - 5 804 116
GenBank mRNA AJ278422.1 - 4 - 5 824 116
AK004972.2 - 4 - 5 804 116
AK128986.1 - 4 - 5 821 116
BC041675.1 - 4 - 5 836 116

View Full Product Details