Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlxipl
Entrez Gene ID
58805
Gene Name
MLX interacting protein-like
Gene Aliases
ChREBP, WS-bHLH, Wbscr14, bHLHd14
Chromosome Location
Chr.5: 135089891 - 135138382 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021455.4 NP_067430.2 1 - 2 324 78
GenBank mRNA AF245475.1 - 1 - 2 302 78
AF245476.1 - 1 - 2 302 78
AF245477.1 - 2 - 3 236 78
AF245478.1 - 1 - 2 302 78
AF245479.1 - 1 - 2 302 78
AK143800.1 - 1 - 2 311 78
AK143999.1 - 1 - 2 325 78
BC064053.1 - 1 - 2 297 78

View Full Product Details