Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ift81
Entrez Gene ID
12589
Gene Name
intraflagellar transport 81
Gene Aliases
AW060663, Cdv-1, Cdv-1r, Cdv1
Chromosome Location
Chr.5: 122540265 - 122614546 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009879.3 NP_034009.2 17 - 18 1902 98
GenBank mRNA AF354757.1 - 17 - 18 1902 98
AK042372.1 - 3 - 4 272 98
BC055721.1 - 17 - 18 1859 98
Y10495.2 - 17 - 18 1902 98
Y10496.1 - 3 - 4 307 98

View Full Product Details