Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pkd2l1
Entrez Gene ID
329064
Gene Name
polycystic kidney disease 2-like 1
Gene Aliases
B830002B15, BC046386, PCL, PKD2L, Pkdl, TRPP3
Chromosome Location
Chr.19: 44147637 - 44248354 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181422.3 NP_852087.2 1 - 2 403 67
GenBank mRNA AB290927.1 - 1 - 2 235 67
AF271381.1 - 1 - 2 369 67
AK046772.1 - 1 - 2 404 67
BC116297.2 - 1 - 2 334 67
BC116323.2 - 1 - 2 334 67

View Full Product Details