The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145501.2 | NP_663476.1 | 8 - 9 | 1345 | 64 |
GenBank mRNA | AK030725.1 | - | 8 - 9 | 1310 | 64 |
BC089309.1 | - | 7 - 8 | 2034 | 64 | |
BC110363.1 | - | 8 - 9 | 1307 | 64 |