Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Abcg3
- Entrez Gene ID
-
27405
- Gene Name
- ATP-binding cassette, sub-family G (WHITE), member 3
- Gene Aliases
-
Abcp2, Mxr2
- Chromosome Location
-
Chr.5: 104935057 - 104982802 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC029095.1 |
- |
6 - 7 |
853 |
62 |
View Full Product Details