Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dgat1
Entrez Gene ID
13350
Gene Name
diacylglycerol O-acyltransferase 1
Gene Aliases
ARAT, C75990, D15Ertd23e, Dgat
Chromosome Location
Chr.15: 76502014 - 76512020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010046.2 NP_034176.1 3 - 4 395 85
GenBank mRNA AB057816.1 - 3 - 4 383 85
AF078752.1 - 3 - 4 453 85
BC003717.1 - 3 - 4 395 85

View Full Product Details