The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010046.2 | NP_034176.1 | 5 - 6 | 537 | 63 |
GenBank mRNA | AB057816.1 | - | 5 - 6 | 525 | 63 |
AF078752.1 | - | 5 - 6 | 595 | 63 | |
BC003717.1 | - | 5 - 6 | 537 | 63 |